ω-Bromo-4-phenylacetophenone 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

146350250 146350050
ACRO146350250EA 98 EUR
ACRO146350250 ACRO146350050
ω-Bromo-4-phenylacetophenone 98%
ω-Bromo-4-phenylacetophenone
Formule: C₁₄H₁₁BrO
Kookpunt: 354 °C (1013 hPa)
Smeltpunt: 123…127 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000202
CAS nummer: 135-73-9
EINECS: 205-217-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Melting point 123 to 127 °C
GC ≥97.5 %
Specific optical rotation -3.0° to -5.0° (20 °C, 589 nm) (c=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR