Pentafluorobenzaldehyde 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

147090025 147090100 147090500
ACRO147090025EA 34.5 EUR
ACRO147090025 ACRO147090100 ACRO147090500
Pentafluorobenzaldehyde 98%
Pentafluorobenzaldehyde
Formule: C₇HF₅O
Kookpunt: 164…166 °C (760 mmHg)
Smeltpunt: 20 °C
Dichtheid: 1,588 g/cm³
Vlampunt: 77 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003303
CAS nummer: 653-37-2
EINECS: 211-502-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR