Cyclohexanol 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

147680010 147680025 147680100
ACRO147680010EA 55.3 EUR
ACRO147680010 ACRO147680025 ACRO147680100
Cyclohexanol 98%
Cyclohexanol
Formule: C₆H₁₂O
Kookpunt: 161 °C (760 mmHg)
Smeltpunt: 23 °C
Dichtheid: 0,96 g/cm³
Vlampunt: 67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003855
CAS nummer: 108-93-0
EINECS: 203-630-6
Merck Index: 13,02754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8878907005758628f5927d7b0f43a99f57de5ae8&vid=c5d64c7b5d58d205d87b42eb97190b1693d3508a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4670 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR