5-Hydroxy-L-tryptophan 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

148290010 148290050 148290250
ACRO148290010EA 45.3 EUR
ACRO148290010 ACRO148290050 ACRO148290250
5-Hydroxy-L-tryptophan 99%
5-Hydroxy-L-tryptophan
Formule: C₁₁H₁₂N₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064341
CAS nummer: 4350-09-8
EINECS: 224-411-1
UN: 2811
ADR: 6.1,III
Merck Index: 12,04895

Order Now

Specification Test Results

Appearance White to light beige to light grey Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Heavy metals ≤10 ppm
Sulfated ash ≤0.3 %
HPLC ≥98.5 % (on dry substance)
Specific optical rotation -30.9° to -34.9° (20°C, 589 nm) (c=1, H2O) (on dry substance)
Titration with HClO4 98.5 to 101.5 % (On dry substance)
Arsenic (As) ≤1 ppm

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR