Hexyl acetate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

148500010 148500250 148500025
ACRO148500010EA 0 EUR
ACRO148500010 ACRO148500250 ACRO148500025
Hexyl acetate 99%
Hexyl acetate
Formule: C₈H₁₆O₂
Kookpunt: 168…170 °C (760 mmHg)
Smeltpunt: –80 °C
Dichtheid: 0,871 g/cm³
Vlampunt: 43 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009524
CAS nummer: 142-92-7
EINECS: 205-572-7
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4110 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.869 to 0.873
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR