Acetaldehyde ammonia trimer 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Aldehyde Ammonia Trimer, 2,4,6-Trimethylhexahydro-1,3,5-triazine Trihydrate, Hexahydro-2,4,6-trimethyl-1,3,5-triazine Trihydrate, 2,4,6-trimethyl-1,3,5-triazinane trihydrate

148530250 148531000 148535000
ACRO148530250EA 34.7 EUR
ACRO148530250 ACRO148531000 ACRO148535000
Acetaldehyde ammonia trimer 98%
Acetaldehyde ammonia trimer

CAS No.: 58052-80-5

Formule: C₆H₂₁N₃O₃
MW: 183.25 g/mol
Kookpunt: 110 °C (760 mmHg)
Smeltpunt: 95…97 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149559
CAS nummer: 58052-80-5
UN: 1841
ADR: 9 III

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR