Dimethyl sulphite 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

148751000
ACRO148751000EA 144 EUR
ACRO148751000
Dimethyl sulphite 99%
Dimethyl sulphite
Formule: C₂H₆O₃S
Kookpunt: 126 °C (1013 hPa)
Dichtheid: 1,29 g/cm³
Vlampunt: 30 °C
MDL Nummer: MFCD00008415
CAS nummer: 616-42-2
EINECS: 210-481-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4085 to 1.4105 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR