2-Chlorophenylhydrazinium chloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

149140050 149140250
ACRO149140050EA 27.4 EUR
ACRO149140050 ACRO149140250
2-Chlorophenylhydrazinium chloride 97%
2-Chlorophenylhydrazinium chloride
Formule: C₆H₈Cl₂N₂
Smeltpunt: 186…188 °C
MDL Nummer: MFCD00012928
CAS nummer: 41052-75-9
EINECS: 255-194-1
UN: 3077

Order Now

Specification Test Results

Appearance White to light pink to tan Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
HPLC ≥96.0 %
TLC ≥96.0 %
Specific optical rotation -2.9° to -3.9° (20°C, 589 nm) (c=1 in DMF)
Residual solvents ≤15.0 % EtOAc


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR