Sulfanilzuur 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

150710010 150711000 150710025 150710100
ACRO150710010EA 91 EUR
ACRO150710010 ACRO150711000 ACRO150710025 ACRO150710100
Sulfanilzuur 99%
Sulfanilzuur
Formule: 4-(H₂N)C₆H₄SO₃H
Kookpunt: 363 °C (1013 hPa)
Smeltpunt: 365 °C
Dichtheid: 1,485 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007886
CAS nummer: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Off-white to beige to blue-grey Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR