Bromocresol purple, pure indicator grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

151330050 151330250 151331400 151330010
ACRO151330050EA 38.7 EUR
ACRO151330050 ACRO151330250 ACRO151331400 ACRO151330010
Bromocresol purple, pure indicator grade
Bromocresol purple
Formule: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Smeltpunt: 245 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011681
CAS nummer: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371

Order Now

Specification Test Results

Appearance Light pink to pink-orange Powder
Infrared spectrum Conforms
Solubility (0.1 % in 50 % ethanol) Clear
UV (20 mg/l in methanol) Complies
Visual transition interval from pH 5.2 (yellow) to pH 6.8 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR