Methyl orange, pure indicator

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

151421000 151425000
ACRO151421000EA 47.4 EUR
ACRO151421000 ACRO151425000
Methyl orange, pure indicator
Methyl orange
Formule: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Kookpunt: ∼90 °C (1013 hPa)
Smeltpunt: >300 °C
Vlampunt: 38
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007502
CAS nummer: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1 III
Merck Index: 13,06127

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
UV Lambda max 461 to 469 nm (in 0.01N NaOH)
Visual transition interval from pH 3.2 (pink or red)to pH 4.4 (yellow-orange)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR