1-Bromo-3-chlorobenzene 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

154200250 154201000
ACRO154200250EA 26.2 EUR
ACRO154200250 ACRO154201000
1-Bromo-3-chlorobenzene 99%
1-Bromo-3-chlorobenzene
Formule: C₆H₄BrCl
Kookpunt: 196 °C (1013 hPa)
Dichtheid: 1,63 g/cm³
Vlampunt: 80 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000568
CAS nummer: 108-37-2
EINECS: 203-575-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5760 to 1.5780 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR