p-Xyleen 99%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

158770010 158770025 158775000 158770100
ACRO158770010EA 47 EUR
ACRO158770010 ACRO158770025 ACRO158775000 ACRO158770100
p-Xyleen 99%, pure
p-Xyleen
Formule: C₆H₄(CH₃)₂
Kookpunt: 138 °C (760 mmHg)
Smeltpunt: 13 °C
Dichtheid: 0,866 g/cm³
Vlampunt: 25 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008556
CAS nummer: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20°C, 589 nm)
GC ≥99.4 %
Residue after evaporation ≤0.005 %
Chloride (Cl) ≤5 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR