Diethyloxalaat 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Oxalic acid diethyl ester

159180010 159180025 159180250 159185000 159180100
ACRO159180010EA 51.3 EUR
ACRO159180010 ACRO159180025 ACRO159180250 ACRO159185000 ACRO159180100
Diethyloxalaat 99%
Diethyloxalaat
Formule: C₆H₁₀O₄
MW: 146.14 g/mol
Kookpunt: 185 °C (760 mmHg)
Smeltpunt: –41 °C
Dichtheid: 1.07 g/cm³
Vlampunt: 76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009119
CAS nummer: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1 III
Merck Index: 13,03152

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4085 to 1.4105 (20°C, 589 nm)
Water ≤0.05 %
Density (20°C) 1.078 to 1.082 kg/l
GC ≥98.5 %
Color scale ≤15 APHA
Acidity ≤0.01 % (as oxalic acid)
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR