Benzyl(dimethyl)amine 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

159751000 159755000 159750025 159750051
ACRO159751000EA 22.8 EUR
ACRO159751000 ACRO159755000 ACRO159750025 ACRO159750051
Benzyl(dimethyl)amine 99%
Benzyl(dimethyl)amine
Formule: C₆H₅CH₂N(CH₃)₂
Kookpunt: 183…184 °C (760 mmHg)
Smeltpunt: –75 °C
Dichtheid: 0,9 g/cm³
Vlampunt: 54 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008329
CAS nummer: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=7044105a7e7fbce7e00f1d15d7a4e017a1b9d418&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5020 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %
Filt.over 0.2 micron PFTE Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR