Methyl vanillate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methyl 4-hydroxy-3-methoxybenzoate

160300050 160300250 160301000
ACRO160300050EA 30.1 EUR
ACRO160300050 ACRO160300250 ACRO160301000
Methyl vanillate 99%
Methyl vanillate
Formule: C₉H₁₀O₄
MW: 182.18 g/mol
Kookpunt: 285…287 °C (760 mmHg)
Smeltpunt: 64…67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008438
CAS nummer: 3943-74-6
EINECS: 223-525-9

Order Now

Specification Test Results

Appearance Light yellow to beige Powder and/or granules
Infrared spectrum Conforms
Melting point 64 to 67 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR