Ethyl crotonate 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester

161381000 161385000
ACRO161381000EA 34.1 EUR
ACRO161381000 ACRO161385000
Ethyl crotonate 96%
Ethyl crotonate
Formule: C₆H₁₀O₂
MW: 114.14 g/mol
Kookpunt: 143
Smeltpunt: –79 °C
Dichtheid: 0.918 g/cm³
Vlampunt: 34 °C
MDL Nummer: MFCD00009289
CAS nummer: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3 II
Merck Index: 13,02625

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4280 (20°C, 589 nm)
Density (20/4°C) 0.916 to 0.921 g/ml
GC ≥95 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR