4,4'-Dipyridyl disulphide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

162240010 162240050 162240250
ACRO162240010EA 40.9 EUR
ACRO162240010 ACRO162240050 ACRO162240250
4,4'-Dipyridyl disulphide 98%
4,4'-Dipyridyl disulphide
Formule: C₁₀H₈N₂S₂
Kookpunt: 348 °C (1013 hPa)
Smeltpunt: 75…79 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006423
CAS nummer: 2645-22-9
EINECS: 220-158-6
UN: 3335

Order Now

Specification Test Results

Appearance White to orange Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 79 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR