2,4-Dichloropyrimidine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

162650250 162650050 162651000
ACRO162650250EA 136 EUR
ACRO162650250 ACRO162650050 ACRO162651000
2,4-Dichloropyrimidine 97%
2,4-Dichloropyrimidine
Formule: C₄H₂Cl₂N₂
Kookpunt: 101 °C (23 mmHg)
Smeltpunt: 57…62 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006061
CAS nummer: 3934-20-1
EINECS: 223-508-6

Order Now

Specification Test Results

Appearance White to yellow to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 57 to 62 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR