5-Methoxysalicylaldehyde 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

164160050 164160100 164160500
ACRO164160050EA 89.5 EUR
ACRO164160050 ACRO164160100 ACRO164160500
5-Methoxysalicylaldehyde 98%
5-Methoxysalicylaldehyde
Formule: C₈H₈O₃
Kookpunt: 103 °C (2,5 mmHg)
Smeltpunt: 4 °C
Dichtheid: 1,21 g/cm³
Vlampunt: >112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003332
CAS nummer: 672-13-9
EINECS: 211-589-0

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR