Tetrahydrofuraan 99.6% stabilised ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Butylene oxide, THF, Diethylene oxide

Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

164240010 164240025 164240050 164240250
ACRO164240010EA 79 EUR
ACRO164240010 ACRO164240025 ACRO164240050 ACRO164240250
Tetrahydrofuraan 99.6% stabilised ACS
Tetrahydrofuraan
Formule: C₄H₈O
MW: 72.11 g/mol
Kookpunt: 66 °C (760 mmHg)
Smeltpunt: –108.4 °C
Dichtheid: 0.88 g/cm³
Vlampunt: –21 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005356
CAS nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Bereik: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Stabilizer 200 to 400 ppm BHT
Residue after ignition ≤0.002 %
Acidity/alkalinity ≤0.0002 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.05 ppm
Peroxides ≤0.0025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR