4,4'-Bis(dimethylamino)benzofenon 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

164490050 164491000 164495000
ACRO164490050EA 23.1 EUR
ACRO164490050 ACRO164491000 ACRO164495000
4,4'-Bis(dimethylamino)benzofenon 98%
4,4'-Bis(dimethylamino)benzofenon
Formule: C₁₇H₂₀N₂O
Kookpunt: 428 °C (1013 hPa)
Smeltpunt: 171…176 °C
Vlampunt: 220 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008312
CAS nummer: 90-94-8
EINECS: 202-027-5
ADR: 6.1,III
Merck Index: 12,06265

Order Now

Specification Test Results

Appearance Blue to dark blue to grey Powder
Infrared spectrum Conforms
Melting point 171 to 176 °C (Dec)
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR