(±)-2-Bromobutyric acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

164510010 164512500 164510050
ACRO164510010EA 338 EUR
ACRO164510010 ACRO164512500 ACRO164510050
(±)-2-Bromobutyric acid 99%
(±)-2-Bromobutyric acid
Formule: H₃CCH₂BrCHCOOH
Kookpunt: 214 °C (1013 hPa)
Smeltpunt: –4 °C
Dichtheid: 1,56 g/cm³
Vlampunt: >112 °C
MDL Nummer: MFCD00004216
CAS nummer: 80-58-0
EINECS: 201-294-5
UN: 2922
ADR: 8,II
Merck Index: 13,01397

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4755 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR