Trifluorazijnzuur-D (99% D) for NMR spectroscopy

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Trifluoroacetic [2H]acid, TFA-D, Trifluoroacetic acid-[²H₁]

166400050 166400100 166400250 166401000
ACRO166400050EA 0 EUR
ACRO166400050 ACRO166400100 ACRO166400250 ACRO166401000
Trifluorazijnzuur-D (99% D) for NMR spectroscopy
Trifluorazijnzuur-D
Formule: F₃CCOOD
Kookpunt: 75 °C (760 mmHg)
Smeltpunt: –15 °C
Dichtheid: 1,5 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00044563
CAS nummer: 599-00-8
EINECS: 209-961-2
UN: 2699
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR