4-Bromo-2-chloroaniline 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

167480100 167480500
ACRO167480100EA 40.9 EUR
ACRO167480100 ACRO167480500
4-Bromo-2-chloroaniline 99%
4-Bromo-2-chloroaniline
Formule: C₆H₅BrClN
Kookpunt: 264 °C (1013 hPa)
Smeltpunt: 67…70 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007660
CAS nummer: 38762-41-3
EINECS: 254-118-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥98.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR