Ammonium formate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Formic acid ammonium salt

168610010 168615000 168610051
ACRO168610010EA 65.6 EUR
ACRO168610010 ACRO168615000 ACRO168610051
Ammonium formate 97%
Ammonium formate
Formule: HCO₂NH₄
Smeltpunt: 115…120 °C
Dichtheid: 1,28 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013103
CAS nummer: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR