Methyl (diethylphosphono)acetate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Diethyl methoxycarbonylmethanephosphonate, Methyl 2-(diethoxyphosphoryl)acetate, Methyl diethylphosphonoacetate

169980100 169980500
ACRO169980100EA 36.6 EUR
ACRO169980100 ACRO169980500
Methyl (diethylphosphono)acetate 97%
Methyl (diethylphosphono)acetate
Formule: C₇H₁₅O₅P
Kookpunt: 127…131 °C (9 mmHg)
Dichtheid: 1,14 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009081
CAS nummer: 1067-74-9
EINECS: 213-938-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4350 (20°C, 589 nm)
GC ≥96.0 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR