2-Furonitrile 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

170430050 170430250
ACRO170430050EA 68.3 EUR
ACRO170430050 ACRO170430250
2-Furonitrile 99%
2-Furonitrile
Formule: C₅H₃NO
Kookpunt: 146 °C (1013 hPa)
Dichtheid: 1,06 g/cm³
Vlampunt: 35 °C
MDL Nummer: MFCD00003223
CAS nummer: 617-90-3
EINECS: 210-537-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear light yellow to light orange Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR