Bis(4-nitrophenyl)carbonate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

171600010 171600050 171600250
ACRO171600010EA 18.3 EUR
ACRO171600010 ACRO171600050 ACRO171600250
Bis(4-nitrophenyl)carbonate 97%
Bis(4-nitrophenyl)carbonate
Formule: C₁₃H₈N₂O₇
Smeltpunt: 139…141 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007322
CAS nummer: 5070-13-3

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 136 to 144 °C
GC ≥96.0 %
Moisture 6 % to 10 %
Bulk density 475 to 625 kg/m³
Acidity ≤0.1 mg KOH/g (Free Acidity)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR