(±)-2-Aminobutyric acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

172091000 172090250
ACRO172091000EA 118 EUR
ACRO172091000 ACRO172090250
(±)-2-Aminobutyric acid 99%
(±)-2-Aminobutyric acid
Formule: C₄H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008093
CAS nummer: 2835-81-6
EINECS: 220-616-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR