DL-Leucine 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

172130250 172132500
ACRO172130250EA 36 EUR
ACRO172130250 ACRO172132500
DL-Leucine 99+%
DL-Leucine
Formule: C₆H₁₃NO₂
Kookpunt: 226 °C (1013 hPa)
Smeltpunt: 293…296 °C
Dichtheid: 1,191 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063087
CAS nummer: 328-39-2
EINECS: 206-328-2
Merck Index: 13,05470

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR