4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

172730010 172730050 172732500
ACRO172730010EA 140 EUR
ACRO172730010 ACRO172730050 ACRO172732500
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)
Formule: C₉H₁₉N₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006479
CAS nummer: 14691-88-4
EINECS: 238-738-2
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Red to red-brown Adhering crystals or needles or moist mass
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR