Diethylether 99.5% stabilised, meets analytical specification of Ph.Eur. for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

176820010 176820025 176820050 176820250
ACRO176820010EA 70.7 EUR
ACRO176820010 ACRO176820025 ACRO176820050 ACRO176820250
Diethylether 99.5% stabilised, meets analytical specification of Ph.Eur. for analysis
Diethylether
Formule: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kookpunt: 34.6 °C (760 mmHg)
Smeltpunt: –116 °C
Dichtheid: 0.714 g/cm³
Vlampunt: –45 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011646
CAS nummer: 60-29-7
UN: 1155
ADR: 3 I
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (K.F.)
GC ≥99.5 %
Color scale ≤10 APHA
Stabilizer 5 ppm BHT
Relative density 0.714 to 0.716 kg/l
Residue after evaporation ≤0.002 %
Acidity passes test
Cadmium (Cd) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.05 ppm
Nickel (Ni) ≤0.02 ppm
Aldehyde (HCHO) ≤0.0001 %
Acetone ≤0.002 %
Ethanol ≤0.01 %
Methanol ≤0.01 %
Peroxides ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR