tert-Butyl hydroperoxide 70%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

180340050 180342500 180345000 180340025 180340100
ACRO180340050EA 21.4 EUR
ACRO180340050 ACRO180342500 ACRO180345000 ACRO180340025 ACRO180340100
tert-Butyl hydroperoxide 70%
tert-Butyl hydroperoxide
Formule: C₄H₁₀O₂
Kookpunt: 96 °C (ontbinding)
Smeltpunt: –3 °C
Dichtheid: 0,937
Vlampunt: 43 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002130
CAS nummer: 75-91-2
EINECS: 200-915-7
UN: 3109
ADR: 5.2,
Merck Index: 14,1570

Order Now

Specification Test Results

Appearance Clear solution
Titration Iodometric 68.5 to 71.0 %
Color scale ≤20 APHA
Titration with HClO4 ≥97.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR