Dimethyl terephthalate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Terephthalic acid dimethyl ester

180580010 180580025 180580250 180585000
ACRO180580010EA 56 EUR
ACRO180580010 ACRO180580025 ACRO180580250 ACRO180585000
Dimethyl terephthalate 99%
Dimethyl terephthalate
Formule: C₁₀H₁₀O₄
MW: 194.19 g/mol
Kookpunt: 288
Smeltpunt: 140…143 °C
Vlampunt: 154 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008440
CAS nummer: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306

Order Now

Specification Test Results

Appearance White Flakes and/or pellets
Infrared spectrum Conforms
Melting point 140 to 143 °C
GC ≥98.5 %
Acid value ≤0.03 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR