Ethyleenglycolmonomethylether (2-Methoxyethanol) 99+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

180790010 180790025 180790250
ACRO180790010EA 68.2 EUR
ACRO180790010 ACRO180790025 ACRO180790250
Ethyleenglycolmonomethylether (2-Methoxyethanol) 99+%, extra pure
Ethyleenglycolmonomethylether (2-Methoxyethanol)
Formule: C₃H₈O₂
Kookpunt: 124,6 °C (1013 hPa)
Smeltpunt: –85 °C
Dichtheid: 0,96 g/cm³
Vlampunt: 38 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002867
CAS nummer: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4040 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR