Benzylchloride 99.5+% stabilised, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (Chloromethyl)benzene, Chlorophenylmethane, α-Chlorotoluene

180850010 180850025 180850100
ACRO180850010EA 43.6 EUR
ACRO180850010 ACRO180850025 ACRO180850100
Benzylchloride 99.5+% stabilised, extra pure
Benzylchloride
Formule: (C₆H₅)CH₂Cl
MW: 126.59 g/mol
Kookpunt: 179.3
Smeltpunt: –39 °C
Dichtheid: 1.1 g/cm³
Vlampunt: 67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000889
CAS nummer: 100-44-7
EINECS: 202-853-6
UN: 1738
ADR: 6.1 II
Merck Index: 13,01131

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5390 (20°C, 589 nm)
GC ≥99.50 %
Color scale ≤20 APHA
Stabilizer 0.25 % propylene oxide

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR