N,N-Dimethylaceetamide 99+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

181070010 181070025 181070050 181070250
ACRO181070010EA 54.2 EUR
ACRO181070010 ACRO181070025 ACRO181070050 ACRO181070250
N,N-Dimethylaceetamide 99+%, extra pure
N,N-Dimethylaceetamide
Formule: CH₃CON(CH₃)₂
Kookpunt: 166,1 °C (1013 hPa)
Smeltpunt: –20 °C
Dichtheid: 0,937 g/cm³
Vlampunt: 70 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008686
CAS nummer: 127-19-5
EINECS: 204-826-4
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.01 %
Free acid (CH3COOH) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR