Dimethyl sulphoxide-[D₆] 99.9 atom % D + 1% TMS NMR

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

181150100 181150250 181150500
ACRO181150100EA 64.7 EUR
ACRO181150100 ACRO181150250 ACRO181150500
Dimethyl sulphoxide-[D₆] 99.9 atom % D + 1% TMS NMR
Dimethyl sulphoxide-[D₆]
Formule: D₃CS(O)CD₃
MW: 84.09 g/mol
Kookpunt: 189
Smeltpunt: 20.2
Dichtheid: 1.19 g/cm³
Vlampunt: 88 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002090
CAS nummer: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.85 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR