Bis(trimethylsilyl)acetylene 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

182010100 182010500
ACRO182010100EA 61.4 EUR
ACRO182010100 ACRO182010500
Bis(trimethylsilyl)acetylene 99%
Bis(trimethylsilyl)acetylene
Formule: C₈H₁₈Si₂
Kookpunt: 136…137 °C (760 mmHg)
Smeltpunt: 21…24 °C
Dichtheid: 0,75 g/cm³
Vlampunt: 2 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008276
CAS nummer: 14630-40-1
EINECS: 238-671-9
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR