15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

182860050 182860250
ACRO182860050EA 40.7 EUR
ACRO182860050 ACRO182860250
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)
Formule: C₁₀H₂₀O₅
Kookpunt: 100…135 °C (,2 mmHg)
Smeltpunt: –20 °C
Dichtheid: 1,1 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005110
CAS nummer: 33100-27-5
EINECS: 251-379-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4670 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥96.0 %
Specific optical rotation +48° to +55° (20°C, 589 nm) (0.5% in Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR