Polyvinyl alcohol, M.W. ∼ 146,000-186,000 98.0-98.8 %, hydrolysed

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: PVOH, PVA, Mowiol®

183120010 183120025 183121000
ACRO183120010EA 156 EUR
ACRO183120010 ACRO183120025 ACRO183121000
Polyvinyl alcohol, M.W. ∼ 146,000-186,000 98.0-98.8 %, hydrolysed
Polyvinyl alcohol
Formule: (CH₂CHOH)n
Smeltpunt: 230…240 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081922
CAS nummer: 9002-89-5
Merck Index: 15,7706

Order Now

Specification Test Results

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 62 to 72 cP (4 % in water)
Degree of hydrolysis 98.0 to 98.8 mol %
Ash ≤1.2 % (as Na2O)
pH 5.0 to 7.0 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR