Pyridinium chlorochromate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

183670250 183671000 183675000
ACRO183670250EA 26.7 EUR
ACRO183670250 ACRO183671000 ACRO183675000
Pyridinium chlorochromate 98%
Pyridinium chlorochromate
Formule: C₅H₆ClCrNO₃
Smeltpunt: 205 °C
MDL Nummer: MFCD00013106
CAS nummer: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08064

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Titration Iodometric 97.5 to 102.5 %
HPLC ≥98.5 %
Residue after ignition ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR