2-Sulphobenzoic cyclic anhydride 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

184750250 184751000
ACRO184750250EA 65.7 EUR
ACRO184750250 ACRO184751000
2-Sulphobenzoic cyclic anhydride 95%
2-Sulphobenzoic cyclic anhydride
Formule: C₇H₄O₄S
Kookpunt: 184…186 °C (18 mmHg)
Smeltpunt: 116…127 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005879
CAS nummer: 81-08-3
EINECS: 201-322-6
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration by coupling ≥94 %
HPLC ≥94 %
Specific optical rotation -17° to -21° (20°C, 589 nm) (0.5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR