Aluminiumoxid basisch Brockmann Grade I 40-300 µm, 60A for chromatography

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

189990010 189990051 189990250
ACRO189990010EA 85.5 EUR
ACRO189990010 ACRO189990051 ACRO189990250
Aluminiumoxid basisch Brockmann Grade I 40-300 µm, 60A for chromatography
Aluminium oxide basic
Formule: Al₂O₃
Kookpunt: 2977 °C (760 mmHg)
Smeltpunt: 2030 °C
Dichtheid: 3,97 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003424
CAS nummer: 1344-28-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size ≤5 % (<40 µm)
Specific surface area 185 to 225 m²/g
pH 8.5 to 10.5 (10% aqueous suspension)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR