Acid green 50, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Lissamine Green B, Green S

190100250
ACRO190100250EA 51.2 EUR
ACRO190100250
Acid green 50, pure
Acid green 50

Appearance: Dark brown to black Powder

Formule: C₂₇H₂₆N₂O₇S₂
MW: 576.61 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012117
CAS nummer: 3087-16-9

Order Now

Specification Test Results

Appearance Dark brown to black Powder
Infrared spectrum Conforms
UV spectrophotometry ≥50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR