Aluminium tri-sec-butanolate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

191215000 191210025 191211000
ACRO191215000EA 107 EUR
ACRO191215000 ACRO191210025 ACRO191211000
Aluminium tri-sec-butanolate 97%
Aluminium tri-sec-butanolate
Formule: C₁₂H₂₇AlO₃
Kookpunt: 200…206 °C (30 mmHg)
Dichtheid: 0,96 g/cm³
Vlampunt: 27 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009327
CAS nummer: 2269-22-9
EINECS: 218-871-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
Assay ≥96.0 % (ex Al)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR