Coomassie® brilliant blue R-250, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

191490050 191490250 191491000 191495000
ACRO191490050EA 26.3 EUR
ACRO191490050 ACRO191490250 ACRO191491000 ACRO191495000
Coomassie® brilliant blue R-250, pure
Coomassie® brilliant blue R-250
Formule: C₄₅H₄₄N₃NaO₇S₂
Smeltpunt: 174…180 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041762
CAS nummer: 6104-59-2
EINECS: 228-060-5

Order Now

Specification Test Results

Appearance Bordeaux to dark purple-brown Crystalline powder
Infrared spectrum Conforms
UV Lambda max 588 to 596 nm (in 50:50 ethanol:water)
Absorption Ratio P-15/P+15 1 to 1.2
Specific absorbance A (1%/1cm) ≥700 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR