Calcium carbonate 99+% for biochemistry

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Marble

192721000 192725000
ACRO192721000EA 40.7 EUR
ACRO192721000 ACRO192725000
Calcium carbonate 99+% for biochemistry
Calcium carbonate
Formule: CaCO₃
Dichtheid: 2,71 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010906
CAS nummer: 471-34-1

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.02 %
Aluminium (Al) ≤0.005 %
Arsenic (As) ≤0.0004 %
Barium (Ba) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.05 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.2 %
Strontium (Sr) ≤0.1 %
Zinc (Zn) ≤0.001 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.01 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR