Iron(III) oxide 99.999% (trace metals basis) -100 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

193260100 193260500 193262500
ACRO193260100EA 74.5 EUR
ACRO193260100 ACRO193260500 ACRO193262500
Iron(III) oxide 99.999% (trace metals basis) -100 mesh
Iron(III) oxide
Formule: Fe₂O₃
Dichtheid: 5,24 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011008
CAS nummer: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now

Specification Test Results

Appearance Red-brown to rust brown Powder
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR